Skip to Content
Merck
All Photos(1)

Key Documents

670049

Sigma-Aldrich

O-[2-(Biotinyl-amino)ethyl]-O′-[3-(N-succinimidyloxy)-3-oxopropyl]polyethylene glycol

Mp 3,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.22

mol wt

Mp 3,000

Ω-end

NHS ester

α-end

biotin

storage temp.

−20°C

Application

O-[2-(Biotinyl-amino)ethyl]-O′-[3-(N-succinimidyloxy)-3-oxopropyl]polyethylene glycol may be used to functionalize far red emitting persistent luminescence nanoparticles (PLNP).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro targeting of avidin-expressing glioma cells with biotinylated persistent luminescence nanoparticles.
Maldiney T, et al.
Bioconjugate Chemistry, 23(3), 472-478 (2012)

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service