Skip to Content
Merck
All Photos(2)

Key Documents

667730

Sigma-Aldrich

3-(2-Furyl)propionic acid

97%

Synonym(s):

2-Furanpropanoic acid, 2-Furanpropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,622.00

R$1,622.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$1,622.00

About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,622.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

56-60 °C

functional group

carboxylic acid

SMILES string

OC(=O)CCc1ccco1

InChI

1S/C7H8O3/c8-7(9)4-3-6-2-1-5-10-6/h1-2,5H,3-4H2,(H,8,9)

InChI key

XLTJXJJMUFDQEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Rong et al.
Toxicological sciences : an official journal of the Society of Toxicology, 173(2), 267-279 (2019-11-20)
Mycophenolic acid (MPA) is commonly prescribed for preventing graft rejection after kidney transplantation. The primary metabolic pathways of MPA are hepatic glucuronidation through UDP-glucuronosyltransferase (UGT) enzymes in the formation of MPA-glucuronide (MPAG, major pathway) and MPA-acyl glucuronide (AcMPAG). p-Cresol, a
Toshihiro Sato et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 17(4), 475-484 (2015-01-13)
Organic anion-transporting polypeptide (OATP) 1B1 and OATP1B3 contribute to hepatic uptake of numerous drugs. Thus, reduced OATP1B1 and OATP1B3 activity in chronic kidney disease (CKD) may have a major impact on the hepatic clearance of drugs. The effect of drug-uremic
Susanne Kirchhof et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 96, 217-225 (2015-08-09)
Eight-armed PEG was functionalized with furyl and maleimide groups (8armPEG20k-Fur and 8armPEG20k-Mal); degradable hydrogels were obtained by cross-linking via Diels-Alder chemistry. To increase the stability to degradation, the macromonomers were modified by introducing a hydrophobic 6-aminohexanoic acid spacer between PEG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service