Skip to Content
Merck
All Photos(1)

Key Documents

667714

Sigma-Aldrich

Borane-d3-THF complex solution

1.0 M in THF, contains 0.005 M N-Isopropyl-N-methyl-tert-butylamine as stabilizer, 97.5 atom % D (mimimum)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BD3 · C4H8O
Molecular Weight:
88.96
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

97.5 atom % D (mimimum)

contains

0.005 M N-Isopropyl-N-methyl-tert-butylamine as stabilizer

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

impurities

<5% ethyl ether

density

0.881 g/mL at 25 °C

mass shift

M+3

storage temp.

2-8°C

SMILES string

[2H]B([2H])[2H].C1CCOC1

InChI

1S/C4H8O.BH3/c1-2-4-5-3-1;/h1-4H2;1H3/i;1D3

InChI key

RMCYTHFAWCWRFA-SVLQTQOOSA-N

Application

THF solution of borane-d3 stabilized with N-isopropyl-N-methyl tert-butylamine used for the preparation of isotopically labeled alcohols.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 2335-2335 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service