Skip to Content
Merck
All Photos(1)

Documents

666335

Sigma-Aldrich

Pd EnCat® Kit I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.22

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

Application

Catalysis Screening Kits

A kit of 5 Pd EnCat® catalysts which are micro-encapsulated palladium acetate catalysts with or without phosphine ligands which facilitate carbon-carbon bond forming processes as well as the removal of palladium metal contamination.

Legal Information

EnCat is a registered trademark of Reaxa Ltd.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 644692Pd EnCat® TOTP30, extent of labeling: 0.4 mmol/g Pd loading, TOTP 0.5, Pd 1.0 1 gSDS

  • 644706Pd EnCat® TPP30, extent of labeling: 0.4 mmol/g Pd loading, TPP 0.8, Pd 1.0 1 gSDS

  • 644714Pd EnCat® 30, extent of labeling: 0.4 mmol/g Pd loading 1 gSDS

  • 644722Pd EnCat® 40, extent of labeling: 0.4 mmol/g Pd loading 1 gSDS

  • 658693Pd(II) EnCat® BINAP30, extent of labeling: 0.39 mmol/g Pd loading, BINAP 0.25, Pd 1.0 1 gSDS

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Sol. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT RE 2 Oral

Target Organs

Liver,Kidney, Nervous system

Storage Class Code

4.1B - Flammable solid hazardous materials

Flash Point(F)

-9.4 - -7.6 °F

Flash Point(C)

-23 - -22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Quan Yu et al.
Chemical communications (Cambridge, England), (6), 678-679 (2003-04-22)
A robust and recyclable palladium catalyst [Pd0EnCat] has been prepared by ligand exchange of polyurea-encapsulated palladium(II) acetate with formic acid, resulting in deposition of Pd(0) in the support material; Pd0EnCat is shown to be a highly efficient transfer hydrogenation catalyst
Steven V Ley et al.
Organic letters, 5(24), 4665-4668 (2003-11-25)
[reaction: see text] Pd nanoparticles (approximately 2 nm in size) microencapsulated in polyurea is an efficient and recyclable catalyst for reductive ring-opening hydrogenolysis of epoxides, using either HCOOH/Et(3)N or H(2) as a hydrogen donor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service