662070
4-(4′-Isopentyloxyphenyl)phenylboronic acid
≥95%
Synonym(s):
4-(4-Isoamyloxyphenyl)phenylboronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
≥95%
form
solid
mp
214-220 °C
SMILES string
CC(C)CCOc1ccc(cc1)-c2ccc(cc2)B(O)O
InChI
1S/C17H21BO3/c1-13(2)11-12-21-17-9-5-15(6-10-17)14-3-7-16(8-4-14)18(19)20/h3-10,13,19-20H,11-12H2,1-2H3
InChI key
DCGWYEOALFPYHU-UHFFFAOYSA-N
Other Notes
Contains varying amounts of anhydride
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service