Skip to Content
Merck
All Photos(1)

Key Documents

655872

Sigma-Aldrich

N-Boc-piperidine

98%

Synonym(s):

1-Boc-piperidine, N-(tert-Butoxycarbonyl)piperidine, N-Boc-tetrahydropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO2
CAS Number:
Molecular Weight:
185.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.454 (lit.)

density

0.964 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CCCCC1

InChI

1S/C10H19NO2/c1-10(2,3)13-9(12)11-7-5-4-6-8-11/h4-8H2,1-3H3

InChI key

RQCNHUCCQJMSRG-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Piperazine derivatives for orally active acetyl-CoA carboxylase 1/2 non-selective inhibitors
  • 2-Aryl and 2-vinyl piperidines
  • Allosteric IGF-1R inhibitors

Reactant for:
  • Allylation and conjugate addition reactions
  • Asymmetric deprotonation
  • Enantioselective inter- and intramolecular aldol reactions
Substrate used in a two-step α-arylation procedure.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron Letters, 47, 455-455 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service