Skip to Content
Merck
All Photos(1)

Key Documents

655171

Sigma-Aldrich

3-Chloro-5-fluorobenzonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$1,399.00

R$1,399.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
R$1,399.00

About This Item

Linear Formula:
ClC6H3(F)CN
CAS Number:
Molecular Weight:
155.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,399.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

54-59 °C (lit.)

functional group

chloro
fluoro
nitrile

SMILES string

Fc1cc(Cl)cc(c1)C#N

InChI

1S/C7H3ClFN/c8-6-1-5(4-10)2-7(9)3-6/h1-3H

InChI key

GIKLRWNRBOLRDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

192.2 °F

Flash Point(C)

89 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service