Skip to Content
Merck
All Photos(1)

Key Documents

655090

Sigma-Aldrich

3-Methyl-1-butylboronic acid

Synonym(s):

Isopentylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13BO2
CAS Number:
Molecular Weight:
115.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

80-91 °C

storage temp.

2-8°C

SMILES string

CC(C)CCB(O)O

InChI

1S/C5H13BO2/c1-5(2)3-4-6(7)8/h5,7-8H,3-4H2,1-2H3

InChI key

UVMWVBMFEDQYRW-UHFFFAOYSA-N

Application

Reactant for:
  • Selective copper-promoted cross-coupling with aromatic amines
  • Synthesis of indazole based analog sensitive Akt inhibitors
  • Preparation of alkylarenes via tetraphosphine-palladium-catalyzed Suzuki cross-coupling with aryl halides

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reuven Rasooly et al.
Journal of food protection, 75(5), 951-954 (2012-05-09)
The potential use of ricin as an agent of biological warfare highlights the need to develop fast and effective methods to detect biologically active ricin. The current "gold standard" for ricin detection is an in vivo mouse bioassay; however, this
Reuven Rasooly et al.
The Journal of biological chemistry, 287(33), 27924-27929 (2012-06-27)
Ricin is a highly toxic protein produced by the castor plant Ricinus communis. The toxin is relatively easy to isolate and can be used as a biological weapon. There is great interest in identifying effective inhibitors for ricin. In this
Reuven Rasooly et al.
Toxins, 5(8), 1447-1461 (2013-08-21)
Aflatoxin-producing fungi contaminate food and feed during pre-harvest, storage and processing periods. Once consumed, aflatoxins (AFs) accumulate in tissues, causing illnesses in animals and humans. Most human exposure to AF seems to be a result of consumption of contaminated plant
Reuven Rasooly et al.
PloS one, 8(8), e71682-e71682 (2013-08-14)
Gene expression in transduced mammalian cells correlates with virus titer, but high doses of vector for gene therapy leads to toxicity in humans and in animals. Changing the optimal tissue culture medium by adding low levels of environmental stressors, such
Lisa J Hill et al.
NPJ Regenerative medicine, 6(1), 3-3 (2021-01-09)
Fibrotic disease is a major cause of mortality worldwide, with fibrosis arising from prolonged inflammation and aberrant extracellular matrix dynamics. Compromised cellular and tissue repair processes following injury, infection, metabolic dysfunction, autoimmune conditions and vascular diseases leave tissues susceptible to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service