Skip to Content
Merck
All Photos(2)

Key Documents

654787

Sigma-Aldrich

2,3′-Dibromoacetophenone

97%

Synonym(s):

α,3′-Dibromoacetophenone, 3-Bromophenacyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$311.00

R$311.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
5 G
R$311.00

About This Item

Linear Formula:
BrC6H4COCH2Br
CAS Number:
Molecular Weight:
277.94
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$311.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

47-52 °C (lit.)

functional group

bromo
ketone

SMILES string

BrCC(=O)c1cccc(Br)c1

InChI

1S/C8H6Br2O/c9-5-8(11)6-2-1-3-7(10)4-6/h1-4H,5H2

InChI key

MZBXSQBQPJWECM-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service