Skip to Content
Merck
All Photos(1)

Key Documents

646652

Sigma-Aldrich

Poly(2,5-di(hexyloxy)cyanoterephthalylidene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C42H58N2O4)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

semiconductor properties

N-type (mobility~10−5 cm2/V·s)

General description

Light emitting conjugated polymer.
Poly(2,5-di(hexyloxy)cyanoterephthalylidene) (CN-PPV) is an electron conducting polymer (n-type) that is a cyano-derivative of poly(phenylenevinylene) (PPV). It can be used in combination with other PPVs for organic photovoltaics.[1][2]

Application

CN-PPV can be used in the formation of conducting films for the fabrication of polymeric light emitting diodes (PLEDs) and photovoltaic cells.[1][2][3][4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of annealing and blending of photoactive polymers on their crystalline structure
Ruderer MA, et al.
The Journal of Physical Chemistry B, 114(47), 15451-15458 (2010)
Optically observed multiple inter-chain interactions in polyblend semiconducting polymer nanoparticles
Hashim Z, et al.
Royal Society of Chemistry Advances, 7(76), 48308-48314 (2017)
Organic light-emitting devices for solid-state lighting
So F, et al.
Mrs Bulletin, 33(7), 663-669 (2008)
Change of photoluminescence properties of cyano-substituted poly p-phenylene vinylene controlled by photoirradiation
Kuzuoka Y and Yamaguchi Y
Synthetic Metals, 158(12), 489-496 (2008)

Articles

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service