Skip to Content
Merck
All Photos(1)

Key Documents

644218

Sigma-Aldrich

Poly(2,5-bis(1,4,7,10-tetraoxaundecyl)-1,4-phenylenevinylene)

Synonym(s):

BTEM-PPV, Poly(2,5-bis(2-(2-(2-methoxyethoxy)ethoxy)ethoxy)-1,4-phenylenevinylene), Poly[2,5-bis(triethoxymethoxy)-1,4-phenylene vinylene]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C22H34O8)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

fluorescence

λex 286 nm; λem 433 nm in chloroform

General description

Poly(2,5-bis(1,4,7,10-tetraoxaundecyl)-1,4-phenylenevinylene) (BTEM-PPV) is a polyphenylenevinylene derivative with good electrical and ionic conductivities. It has a PPV as the backbone and the side chains consist of oligo(ethylene oxide) that can be utilized for transporting of ions. It can be used in the fabrication of organic electronic based devices.

Application

BTEM-PPV is a conjugating polymer that can be used in the fabrication of red-light emitting electrochemical cells (LECs). These LECs can potentially be used in the formation of novel chemical sensor systems and organic light emitting diodes (OLEDs).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Light-emitting electrochemical cells and light-emitting diodes based on ionic conductive poly (phenylene vinylene): a new chemical sensor system.
Holzer L, et al.
Synthetic Metals, 100(1), 71-77 (1999)
Novel poly (p-phenylene vinylene) derivatives with oligo (ethylene oxide) side chains: synthesis and pattern formation.
Winkler B, et al.
Chemistry of Materials, 11(3), 704-711 (1999)
Organic-inorganic hybrid light-emitting composites: Poly (p-phenylene vinylene) intercalated clay nanoparticles.
Winkler B, et al.
Journal of Materials Science Letters, 18(19), 1539-1541 (1999)
New luminescent polymers for LEDs.
Chuah BS, et al.
Synthetic Metals, 91(1-3), 279-282 (1997)
Ionochromism in a light-emitting electrochemical cell with low response time based on an ionic conductive poly-phenylene vinylene.
Holzer L, et al.
Applied Physics Letters, 75(14), 2014-2016 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service