Skip to Content
Merck
All Photos(2)

Key Documents

636401

Sigma-Aldrich

Phthalylglycyl chloride

96%

Synonym(s):

1,3-Dioxo-2-isoindolineacetyl chloride, NSC 401729, Phthalylglycine acid chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$641.00

R$641.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$641.00

About This Item

Empirical Formula (Hill Notation):
C10H6ClNO3
CAS Number:
Molecular Weight:
223.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$641.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

functional group

acyl chloride
imide

SMILES string

ClC(=O)CN1C(=O)c2ccccc2C1=O

InChI

1S/C10H6ClNO3/c11-8(13)5-12-9(14)6-3-1-2-4-7(6)10(12)15/h1-4H,5H2

InChI key

RHZBRCQIKQUQHQ-UHFFFAOYSA-N

General description

Phthalylglycyl chloride can be synthesized via reaction of phthalyl-glycine with thionyl chloride.[1]

Application

Phthalylglycyl chloride can be used in the synthesis of N-Phthalimidoacetyl-β-phenylethylamnine.[2]
Reactant involved in:
  • Synthesis of selenium-containing amino acid analogs with bactericidal and antifungal activity
  • Photoinduced single electron transfer cyclization for preparation of cyclic peptides
  • Arndt-Eistert synthesis
  • Mitsunobu and arbuzov-type multicomponent pathway
  • Remote C-H bond functionalization
  • Amido-alkylation for synthesis of benzofuran derivatives

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Enantioselective synthesis of (R)-(-)-praziquantel (PZQ)"
Roszkowski P, et al.
Tetrahedron Asymmetry, 17(09), 1415-1419 (2006)
"Synthesis and stereochemistry of azeto [2, 1-a] isoquinolin-2-one derivatives"
Csomos P,et al.
ARKIVOC (Gainesville, FL, United States), 87, 93-93 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service