Skip to Content
Merck
All Photos(1)

Key Documents

634808

Sigma-Aldrich

1,2,3,5-Tetra-O-acetyl-α-D-arabinofuranose

technical grade

Synonym(s):

NSC 108078

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O9
CAS Number:
Molecular Weight:
318.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

≥80%

refractive index

n20/D 1.4500 (lit.)

bp

140 °C/0.01 mmHg (lit.)

density

1.31 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)OC[C@H]1O[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C13H18O9/c1-6(14)18-5-10-11(19-7(2)15)12(20-8(3)16)13(22-10)21-9(4)17/h10-13H,5H2,1-4H3/t10-,11-,12+,13+/m1/s1

InChI key

IHNHAHWGVLXCCI-NDBYEHHHSA-N

Application

Reactant for:
  • Asymmetric synthesis of hydroxyl acetylpentofuranosides via diastereoselective enzymic deacetylation through Candida antarctica B lipase (CAL B)-catalyzed alcoholysis
  • Reaction with trimethylaluminum in synthesis of 1,2-O-isopropylidene D-ribofuranose
  • Synthesis of oligosaccharides of motifs D and E of arabinogalactan present in Mycobacterium tuberculosis via coupling and glycosylation reactions

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service