Skip to Content
Merck
All Photos(1)

Key Documents

634379

Sigma-Aldrich

2-Keto-3-(methyl-13C,d2)-butyric acid-4-13C,d2 sodium salt

98 atom % D, 98 atom % 13C

Synonym(s):

α-Keto-3-(methyl-13C,d2)-butyric acid-4-13C,d2 sodium, α-Ketoisovaleric acid-13C2,d4 sodium salt, 3-(Methyl-13C,d2)-2-oxobutanoic-4-13C,d2 acid sodium salt, Sodium α-ketoisovalerate-3-(methyl-13C,d2),4-13C,d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD2CH(13CD2H)COCO2Na
Molecular Weight:
144.11
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % 13C
98 atom % D

form

solid

technique(s)

bio NMR: suitable

mp

227-230 °C (lit.)

mass shift

M+6

SMILES string

[Na+].[2H][13CH]([2H])C([13CH]([2H])[2H])C(=O)C([O-])=O

InChI

1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1/i1+1D2,2+1D2;

InChI key

WIQBZDCJCRFGKA-YVAKGFCFSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service