Skip to Content
Merck
All Photos(1)

Documents

634050

Sigma-Aldrich

5-Bromo-1-(tert-butyldimethylsilyl)-1H-indole

97%

Synonym(s):

5-Bromo-1-TBDMS-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20BrNSi
CAS Number:
Molecular Weight:
310.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.574

bp

158 °C/0.10 mmHg

density

1.224 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)n1ccc2cc(Br)ccc12

InChI

1S/C14H20BrNSi/c1-14(2,3)17(4,5)16-9-8-11-10-12(15)6-7-13(11)16/h6-10H,1-5H3

InChI key

WBOWSDFSONZNEE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service