617512
Dimethyl ether-13C2
99 atom % 13C
Synonym(s):
Methyl ether-13C2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(13CH3)2O
CAS Number:
Molecular Weight:
48.05
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
form
gas
mol wt
48.04 by atom % calculation
mass shift
M+2
SMILES string
[13CH3]O[13CH3]
InChI
1S/C2H6O/c1-3-2/h1-2H3/i1+1,2+1
InChI key
LCGLNKUTAGEVQW-ZDOIIHCHSA-N
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1A - Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
WGK 1
Flash Point(F)
-41.8 °F - closed cup
Flash Point(C)
-41 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service