Skip to Content
Merck
All Photos(1)

Documents

617245

Sigma-Aldrich

1-Bromo-4-fluorobenzene-d4

98 atom % D, 98% (CP)

Synonym(s):

4-Bromofluorobenzene-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6D4F
CAS Number:
Molecular Weight:
179.02
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

bp

150 °C (lit.)

mp

-16 °C (lit.)

density

1.629 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]c1c([2H])c(Br)c([2H])c([2H])c1F

InChI

1S/C6H4BrF/c7-5-1-3-6(8)4-2-5/h1-4H/i1D,2D,3D,4D

InChI key

AITNMTXHTIIIBB-RHQRLBAQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service