Skip to Content
Merck
All Photos(1)

Key Documents

614122

Sigma-Aldrich

Chenodeoxycholic acid-2,2,4,4-d4

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$15,819.00

R$15,819.00


Check Cart for Availability

Request more information

Select a Size

Change View
500 MG
R$15,819.00

About This Item

Empirical Formula (Hill Notation):
C24D4H36O4
Molecular Weight:
396.60
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

R$15,819.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

technique(s)

mass spectrometry (MS): suitable

mp

165-167 °C (lit.)

mass shift

M+4

SMILES string

[2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3[C@H](O)C[C@@H]2C([2H])([2H])[C@@H]1O)[C@H](C)CCC(O)=O

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-/m1/s1/i8D2,12D2

InChI key

RUDATBOHQWOJDD-PSTGXAJBSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sidhartha R Sinha et al.
Cell host & microbe, 27(4), 659-670 (2020-02-27)
Secondary bile acids (SBAs) are derived from primary bile acids (PBAs) in a process reliant on biosynthetic capabilities possessed by few microbes. To evaluate the role of BAs in intestinal inflammation, we performed metabolomic, microbiome, metagenomic, and transcriptomic profiling of
Bin Zan et al.
Biomedical chromatography : BMC, 34(7), e4835-e4835 (2020-03-22)
Bear bile is a valuable medicinal material used in traditional Chinese medicine for over 2000 years. However, developing a substitute has become necessary because of protection measures for this endangered species. The ingredients of in vitro cultured bear bile powder (CBBP)
Susan A Joyce et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(20), 7421-7426 (2014-05-07)
Alterations in the gastrointestinal microbiota have been implicated in obesity in mice and humans, but the key microbial functions influencing host energy metabolism and adiposity remain to be determined. Despite an increased understanding of the genetic content of the gastrointestinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service