613479
Ether-d10
99 atom % D
Synonym(s):
Di(ethyl-d5) ether, Diethyl ether-d10
About This Item
Recommended Products
isotopic purity
99 atom % D
Assay
99% (CP)
form
liquid
expl. lim.
0.34-6.3 % (lit.)
technique(s)
NMR: suitable
bp
33-34 °C (lit.)
mp
-116 °C (lit.)
density
0.801 g/mL at 25 °C
mass shift
M+10
SMILES string
[2H]C([2H])([2H])C([2H])([2H])OC([2H])([2H])C([2H])([2H])[2H]
InChI
1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
InChI key
RTZKZFJDLAIYFH-MWUKXHIBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-40.0 °F - closed cup
Flash Point(C)
-40 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service