All Photos(1)
About This Item
Linear Formula:
(C6H5O)213CO
Molecular Weight:
215.21
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
bp
301-302 °C (lit.)
mp
79-82 °C (lit.)
mass shift
M+1
SMILES string
O=[13C](Oc1ccccc1)Oc2ccccc2
InChI
1S/C13H10O3/c14-13(15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H/i13+1
InChI key
ROORDVPLFPIABK-KCKQSJSWSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service