Skip to Content
Merck
All Photos(1)

Key Documents

606294

Sigma-Aldrich

Ethanolamine-13C2

99 atom % 13C

Synonym(s):

2-Aminoethanol-13C2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$3,277.00

R$3,277.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
R$3,277.00

About This Item

Linear Formula:
HO13CH213CH2NH2
CAS Number:
Molecular Weight:
63.07
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

R$3,277.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

bp

170 °C (lit.)

mp

11 °C (lit.)

density

1.045 g/mL at 25 °C

mass shift

M+2

SMILES string

N[13CH2][13CH2]O

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2/i1+1,2+1

InChI key

HZAXFHJVJLSVMW-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup - (in analogy to similar products)

Flash Point(C)

91 °C - Pensky-Martens closed cup - (in analogy to similar products)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service