Skip to Content
Merck
All Photos(1)

Key Documents

60352

Sigma-Aldrich

Pyrophosphoric acid

≥95% H4P2O7 basis

Synonym(s):

Diphosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2P(O)OP(O)(OH)2
CAS Number:
Molecular Weight:
177.98
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

Assay

≥95% H4P2O7 basis

impurities

≤0.001% heavy metals (as Pb)

cation traces

Fe: ≤20 mg/kg
Na: ≤20 mg/kg

SMILES string

OP(O)(=O)OP(O)(O)=O

InChI

1S/H4O7P2/c1-8(2,3)7-9(4,5)6/h(H2,1,2,3)(H2,4,5,6)

InChI key

XPPKVPWEQAFLFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan H Powdrill et al.
Antimicrobial agents and chemotherapy, 54(3), 977-983 (2009-12-24)
We studied the biochemical mechanisms associated with inhibition and resistance to a 4,5-dihydroxypyrimidine carboxylate that inhibits the hepatitis C virus (HCV) RNA-dependent RNA polymerase NS5B. On the basis of the structure of the pharmacophore, it has been suggested that these
Bruno Marchand et al.
The Journal of biological chemistry, 282(5), 3337-3346 (2006-12-06)
The pyrophosphate (PPi) analogue phosphonoformic acid (PFA or foscarnet) inhibits the reverse transcriptase (RT) of the human immunodeficiency virus type 1 (HIV-1); however, the mechanisms of drug action and resistance remain elusive. Here we studied the effects of the translocational
Tommi Kajander et al.
FEBS letters, 587(13), 1863-1869 (2013-05-21)
Soluble inorganic pyrophosphatases (PPases) catalyse an essential reaction, the hydrolysis of pyrophosphate to inorganic phosphate. In addition, an evolutionarily ancient family of membrane-integral pyrophosphatases couple this hydrolysis to Na(+) and/or H(+) pumping, and so recycle some of the free energy
Stephen J Butler et al.
Chemistry, an Asian journal, 7(11), 2621-2628 (2012-09-12)
The anion binding ability of a family of bis(Zn(II)-Dpa) functionalized cyclic peptides has been investigated using displacement assays with a fluorescent coumarin indicator in water, saline solution, and Krebs buffer. Non-binding side-chain steric bulk, the relative position of binding sites
Rakesh Kumar Pathak et al.
Inorganic chemistry, 51(9), 4994-5005 (2012-04-24)
Pyridyl-based triazole-linked calix[4]arene conjugates, viz. L(1) and L(2), were synthesized and characterized. These two conjugates were shown to be selective and sensitive for Zn(2+) among the 12 metal ions studied in HEPES buffer medium by fluorescence, absorption, and visual color

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service