Assay
95%
mp
78-82 °C (lit.)
SMILES string
C1CCN(C1)B(B(N2CCCC2)N3CCCC3)N4CCCC4
InChI
1S/C16H32B2N4/c1-2-10-19(9-1)17(20-11-3-4-12-20)18(21-13-5-6-14-21)22-15-7-8-16-22/h1-16H2
InChI key
SSJAPCOUEXPQNP-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service