Skip to Content
Merck
All Photos(1)

Key Documents

592293

Sigma-Aldrich

2-Methoxyethyl chloroformate

Synonym(s):

(2-Methoxyethoxy)carbonyl chloride, (Chloro)(2-methoxyethoxy)methanone, 2-Methoxyethyl carbonochloridate, Chloroformic acid 2-methoxyethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCOCl
CAS Number:
Molecular Weight:
138.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.4200 (lit.)

bp

157-158 °C (lit.)

density

1.192 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COCCOC(Cl)=O

InChI

1S/C4H7ClO3/c1-7-2-3-8-4(5)6/h2-3H2,1H3

InChI key

NDYYWMXJZWHRLZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of N-bridged bicyclic sulfonamides as inhibitors of γ-secretase
  • Preparation of dimethoxyethyl azodicarboxylate via amidation/oxidation and application to Mitsunobu reaction
  • Regioselective, stereoselective, and kinetically-controlled nickel-catalyzed cyanoesterification of allenes chloroformates and TMSCN
  • Preparation of amino carboxamidobenzothiazoles as Lck inhibitors
  • Preparation of nonracemic spirooxindoles using a stereoselective three-component coupling reaction as the key step and using acylation reactions to add structural diversity

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.0 °F - closed cup

Flash Point(C)

58.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service