Skip to Content
Merck
All Photos(1)

Documents

579998

Sigma-Aldrich

Poly(Disperse Red 1 acrylate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

transition temp

Tg 79 °C (onset)

solubility

chloroform: soluble

λmax

464

General description

NLO polymer

Application

Poly(Disperse Red 1 acrylate) can be used as a gold standard for surface relief gratings (SRGs). It is a pH responsive solvatochromic dye that can be used in the development of dual sensors for temperature and pH detection.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble polymeric dual sensor for temperature and pH value
Pietsch C, et al.
Angewandte Chemie (International Edition in English), 48(31), 5653-5656 (2009)
Effect of head group size on the photoswitching applications of azobenzene Disperse Red 1 analogues
Goulet-Hanssens A, et al.
Journal of Material Chemistry C, 2(36), 7505-7512 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service