Skip to Content
Merck
All Photos(1)

Key Documents

579858

Sigma-Aldrich

Ethyl 5-fluoroindole-2-carboxylate

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$743.00

R$743.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$743.00

About This Item

Empirical Formula (Hill Notation):
C11H10FNO2
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$743.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

mp

146-150 °C

functional group

ester
fluoro

SMILES string

CCOC(=O)c1cc2cc(F)ccc2[nH]1

InChI

1S/C11H10FNO2/c1-2-15-11(14)10-6-7-5-8(12)3-4-9(7)13-10/h3-6,13H,2H2,1H3

InChI key

VIKOQTQMWBKMNA-UHFFFAOYSA-N

Application

  • Reactant for preparation of antitumor agents
  • Reactant for preparation of antihyperlipidemic agents
  • Reactant for preparation of factor Xa inhibitors
  • Reactant for preparation of potential antiarrhythmic agents
  • Reactant for preparation of inhibitors of the cytosolic phospholipase A2
  • Reactant for preparation of thromboxane receptor antagonist and thromboxane synthase inhibitor via Mitsunobu reaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service