Skip to Content
Merck
All Photos(1)

Key Documents

576700

Sigma-Aldrich

2,2′-Bithiophene-5-carboxaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6OS2
CAS Number:
Molecular Weight:
194.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

55-58 °C (lit.)

SMILES string

[H]C(=O)c1ccc(s1)-c2cccs2

InChI

1S/C9H6OS2/c10-6-7-3-4-9(12-7)8-2-1-5-11-8/h1-6H

InChI key

FYBWRAXKYXTOQC-UHFFFAOYSA-N

Application

2,2′-Bithiophene-5-carboxaldehyde may be used in the synthesis of the following:
  • boron dipyrromethene(BODIPY)oligothiophenes via a multi-step reaction process
  • (2,2′-bithiophene-5-carbaldehyde)-4-nitrophenylhydrazone(BT-NPH) via reaction with 4-nitrophenylhydrazine
  • bithiophene fulleropyrrolidine obtained via refluxing with sarcosine and fullerene
  • azomethine phthalic diimides by heating with N,N-bis(4-amino-2,3,5,6-tetramethylphenyl)naphthalene-1,4,5,8-dicarboximide (DANDI)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Optical nonlinearities and molecular conformations in thiophene-based hydrazone crystals"
Kwon P-O, et al.
The Journal of Physical Chemistry C, 113(34), 15405-15411 (2009)
"The effect of thiophene substituents of fulleropyrrolidine acceptors on the performance of inverted organic solar cells"
Kaunisto.MK, et al.
Synthetic Metals, 195, 193-200 (2014)
"New low band gap compounds comprised of naphthalene diimide and imine units"
Schab-Balcerzak E, et al.
Synthetic Metals, 162(05), 543- 553 (2012)
"Enhanced Functionality for Donor?Acceptor Oligothiophenes by means of Inclusion of Bodipy: Synthesis, Electrochemistry, Photophysics, and Model Chemistry"
Collado D, et al.
Chemistry?A European Journal , 17(02), 498-507 (2011)
Sara S M Fernandes et al.
ACS omega, 3(10), 12893-12904 (2018-11-10)
A series of push-pull heterocyclic N,N-diphenylhydrazones were prepared to study the effect of structural modifications (different π-spacers and electron-withdrawing groups) on the optical (linear and nonlinear) and electronic properties of the molecules. The photovoltaic response of dye-sensitized solar cells assembled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service