Skip to Content
Merck
All Photos(1)

Key Documents

572829

Sigma-Aldrich

3-Hydroxy-4-(hydroxymethyl)-5-(hydroxymethyl-d2)-2-methylpyridine

98 atom % D, 98% (CP)

Synonym(s):

Pyridoxine-(5-hydroxymethyl-d2), Pyridoxol-(5-hydroxymethyl-d2), Vitamin B6-(5-hydroxymethyl-d2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9D2NO3
CAS Number:
Molecular Weight:
171.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

mp

150-151 °C (lit.)

mass shift

M+2

storage temp.

−20°C

SMILES string

[2H]C([2H])(O)c1cnc(C)c(O)c1CO

InChI

1S/C8H11NO3/c1-5-8(12)7(4-11)6(3-10)2-9-5/h2,10-12H,3-4H2,1H3/i3D2

InChI key

LXNHXLLTXMVWPM-SMZGMGDZSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service