Skip to Content
Merck
All Photos(2)

Key Documents

570338

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(ε−caprolactone) methyl ether

PEG average Mn ~5,000, PCL average Mn ~32,000

Synonym(s):

Poly(ethylene glycol) methyl ether-block-poly(ε−caprolactone)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$2,920.00

R$2,920.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$2,920.00

About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

R$2,920.00


Check Cart for Availability

Request a Bulk Order

form

solid

mol wt

PCL average Mn ~32,000
PEG average Mn ~5,000
average Mn ~37,000 (total)

degradation timeframe

>12 months

mp

54.2-55.9 °C (lit.)
54.2-55.9 °C

Mw/Mn

<1.4

PDI

<1.4

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Biocompatible diblock copolymer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service