Skip to Content
Merck
All Photos(1)

Key Documents

565490

Sigma-Aldrich

2-Aminobenzhydrazide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CONHNH2
CAS Number:
Molecular Weight:
151.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

121-125 °C (lit.)

SMILES string

NNC(=O)c1ccccc1N

InChI

1S/C7H9N3O/c8-6-4-2-1-3-5(6)7(11)10-9/h1-4H,8-9H2,(H,10,11)

InChI key

GRWMSCBKWMQPON-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Li et al.
Developmental and comparative immunology, 88, 206-212 (2018-07-27)
Giardia lamblia is one of the most prevalent parasites residing in the duodenum of human and many other mammals throughout the world which is transmitted via ingested cysts through contaminated food or water. The severity of disease may depend on
Zuleyha Almaz et al.
Biotechnology and applied biochemistry, 68(1), 102-113 (2020-02-16)
We have developed efficient procedure for isolation of horseradish peroxidase (HRP) using aminobenzohydrazide-based affinity chromatography. Sepharose 4B-bounded aminobenzohydrazides are suitable for long-term use and large-scale purification. In this study, 26 aminobenzohydrazide derivatives were synthesized, characterized and defined as new HRP
Anand Thangaraj et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 18(6), 1533-1539 (2019-04-18)
A new multi-analyte selective fluorescence chemosensor DA was synthesized by a simple one pot reaction between dansyl chloride and 2-aminobenzohydrazide in the presence of a base. In DMSO : H2O (1 : 1, v/v), the fluorescence of DA at 483 nm was blue shifted
Fatih Inci et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(9), 1800121-1800121 (2018-09-27)
One out of every six American women has been the victim of a sexual assault in their lifetime. However, the DNA casework backlog continues to increase outpacing the nation's capacity since DNA evidence processing in sexual assault casework remains a
Yunjia Zhang et al.
Redox biology, 29, 101409-101409 (2020-01-14)
Atherosclerosis is a chronic inflammatory disease of the vasculature characterised by the infiltration of activated neutrophils and macrophages at sites of damage within the vessel wall, which contributes to lesion formation and plaque progression. Selenomethionine (SeMet) is an organic form

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service