Skip to Content
Merck
All Photos(1)

Key Documents

562858

Sigma-Aldrich

Dibenzofuran-4-carboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8O2
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

94-98 °F (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1cccc2c3ccccc3oc12

InChI

1S/C13H8O2/c14-8-9-4-3-6-11-10-5-1-2-7-12(10)15-13(9)11/h1-8H

InChI key

GQYTWBPRZFRASB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Fitzgerald et al.
Acta crystallographica. Section C, Crystal structure communications, 47 ( Pt 10), 2134-2137 (1991-10-15)
Mr = 196.21, monoclinic, P2(1)/n, a = 6.589 (1), b = 9.784 (2), c = 14.394 (2) A, beta = 95.69 (1) degrees, V = 923.4 (3) A3, Z = 4, Dx = 1.41 g cm-3, lambda(Mo K alpha) =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service