Skip to Content
Merck
All Photos(1)

Key Documents

559199

Sigma-Aldrich

Ethyl (4-iodobenzoyl)acetate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
318.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.6070 (lit.)

density

1.609 g/mL at 25 °C (lit.)

functional group

ester
iodo
ketone

SMILES string

CCOC(=O)CC(=O)c1ccc(I)cc1

InChI

1S/C11H11IO3/c1-2-15-11(14)7-10(13)8-3-5-9(12)6-4-8/h3-6H,2,7H2,1H3

InChI key

DOIUIJYPQHAHLM-UHFFFAOYSA-N

Application

Reactant for:
  • Synthesis of trisubstituted pyrroles via rearrangement reactions catalyzed by Ir catalyst
  • Preparation of biologically active molecules

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service