Skip to Content
Merck
All Photos(1)

Key Documents

558605

Sigma-Aldrich

3,5-Difluoroiodobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3I
CAS Number:
Molecular Weight:
239.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.5540 (lit.)

bp

173-174 °C (lit.)

SMILES string

Fc1cc(F)cc(I)c1

InChI

1S/C6H3F2I/c7-4-1-5(8)3-6(9)2-4/h1-3H

InChI key

QQCFOQNFPIAENW-UHFFFAOYSA-N

General description

3,5-Difluoroiodobenzene is an asymmetric top molecule. 3,5-Difluoroiodobenzene reacts with magnesium turnings in the presence of a minimum amount of ether to afford the corresponding Grignard reagent. This reagent is employed in the synthesis of tris(3,5-difluorophenyl)methanol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

180.0 °F - closed cup

Flash Point(C)

82.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzene solvent induced shifts of the proton magnetic resonance spectra of some benzene derivatives. Importance of charge effects.
Wasylishen R, et al.
Canadian Journal of Chemistry, 48(18), 2885-2895 (1970)
Controlling the rotation of asymmetric top molecules by the combination of a long and a short laser pulse.
Viftrup SS, et al.
Physical Review A: General Physics, 79(2), 023404-023404 (2009)
Perfluoroaromatics. Tris (polyfluoroaryl) carbocations.
Kulkarni SV, et al.
Journal of the American Chemical Society, 95(6), 1859-1864 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service