Skip to Content
Merck
All Photos(1)

Key Documents

557412

Sigma-Aldrich

6-Fluoro-2H-1,4-benzoxazin-3(4H)-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6FNO2
CAS Number:
Molecular Weight:
167.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

207-211 °C (lit.)

functional group

fluoro

SMILES string

Fc1ccc2OCC(=O)Nc2c1

InChI

1S/C8H6FNO2/c9-5-1-2-7-6(3-5)10-8(11)4-12-7/h1-3H,4H2,(H,10,11)

InChI key

ZKNRUFWEZPGGQP-UHFFFAOYSA-N

General description

Due to the presence of halogen (F) at C-6 position in 6-fluoro-2H-1,4-benzoxazin-3(4H)-one (in active form), it exhibits phytotoxic activity.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco A Macías et al.
Journal of agricultural and food chemistry, 54(26), 9843-9851 (2006-12-21)
The utility of benzoxazinones and some of their synthetic derivatives in the search for new leads for herbicide model development has been widely discussed. As the benzoxazinone skeleton contains three different potential areas for functionalization (C-2, N-4, and aromatic protons
Catalytic hydrogenation of sulfur-containing nitrobenzene over Pd/C catalysts: In situ sulfidation of Pd/C for the preparation of Pd x S y catalysts.
Zhang Q, et al.
Applied Catalysis A: General, 497, 17-21 (2015)
Photoelectrochemical reduction of meta-halonitrobenzenes and related species.
Robert AW and George WJ.
J. Chem. Soc. Perkin Trans. II, 8, 1673-1677 (1995)
H B Dunford
Redox report : communications in free radical research, 5(4), 169-171 (2000-09-20)
The first complete mechanistic analysis of halide ion oxidation by a peroxidase was that of iodide oxidation by horseradish peroxidase. It was shown conclusively that a two-electron oxidation of iodide by compound I was occurring. This implied that oxygen atom
The proximate coupling constant, 5 J (H, CH3), and the torsional mobility of the thiomethyl group in some thioanisole derivatives.
Schaefer T, et al.
Canadian Journal of Chemistry, 69(4), 620-624 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service