Skip to Content
Merck
All Photos(1)

Key Documents

556319

Sigma-Aldrich

2-Chloro-5-fluorobenzyl bromide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CH2Br
CAS Number:
Molecular Weight:
223.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.566 (lit.)

bp

65-66 °C/2 mmHg (lit.)

density

1.654 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Cl)c(CBr)c1

InChI

1S/C7H5BrClF/c8-4-5-3-6(10)1-2-7(5)9/h1-3H,4H2

InChI key

AUVLFQDKJFSFIX-UHFFFAOYSA-N

General description

2-Chloro-5-fluorobenzyl bromide can be obtained by reacting 2-chloro-5-fluorotoluene with N-bromosuccinimide.[1]

Application

2-Chloro-5-fluorobenzyl bromide may be used to synthesize 2-chloro-5-fluorobenzaldehyde.[1]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.0 °F - closed cup

Flash Point(C)

17.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Unusual Fluorine Atom Rearrangement in the Photocyclization of 1-Fluoro[ 5]helicenes.
Mallory FB and Mallory CW.
The Journal of Organic Chemistry, 48(4), 526-526 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service