549894
2-Chloro-9-(3-(dimethylamino)propyl)-thioxanthen-9-ol
96%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H20ClNOS
CAS Number:
Molecular Weight:
333.88
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
96%
mp
148-151 °C (lit.)
SMILES string
CN(C)CCCC1(O)c2ccccc2Sc3ccc(Cl)cc13
InChI
1S/C18H20ClNOS/c1-20(2)11-5-10-18(21)14-6-3-4-7-16(14)22-17-9-8-13(19)12-15(17)18/h3-4,6-9,12,21H,5,10-11H2,1-2H3
InChI key
YJINNJOMTOJZBH-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phospholipid-sensitive calcium-dependent protein kinase: inhibition by antipsychotic drugs.
Schatzman RC, et al.
Biochemical and Biophysical Research Communications, 98(3), 669-676 (1981)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service