All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H8BrNOS
CAS Number:
Molecular Weight:
210.09
Beilstein:
6246836
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Assay
≥97.0%
mp
147-149 °C
SMILES string
[Br-].OCC[n+]1ccsc1
Other Notes
Catalyst for the addition of aldehydes to α,β-unsaturated esters (Stetter-Michael-addition)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
L. Novak et al.
Chemische Berichte, 117, 2939-2939 (1980)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service