Skip to Content
Merck
All Photos(1)

Key Documents

543969

Sigma-Aldrich

Triethylamine

99.5%

Synonym(s):

N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

51.75 mmHg ( 20 °C)

Assay

99.5%

autoignition temp.

593 °F

expl. lim.

8 %

availability

available only in EU

refractive index

n20/D 1.401 (lit.)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

solubility

water: soluble 112 g/L at 20 °C

density

0.726 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylamine (TEA, Et3N) is an aliphatic amine. Its addition to matrix-assisted laser desorption/ionization (MALDI) matrices faffords transparent liquid matrices with enhanced ability for spatial resolution during MALDI mass spectrometric (MS) imaging. A head-space gas chromatography (GC) procedure for the determination of triethylamine in active pharmaceutical ingredients has been reported. The viscosity coefficient of triethylamine vapor over a range of density and temperature has been measured.

Application

Triethylamine has been used during grafting of 5,6-epoxyhexyltriethoxysilane on a silicon device.

Legal Information

Product of Arkema Inc.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

GC quantification of cyclopropylamine, diethylamine and triethylamine in active pharmaceutical ingredients.
Raghuram P, et al.
Chromatographia, 71(9-10), 963-966 (2010)
Transparent triethylamine-containing MALDI matrices.
Rubakhin SS and Sweedler JV.
Israel J. Chem., 47(2), 185-193 (2007)
Enhancing surface activity in silicon microreactors: use of black silicon and alumina as catalyst supports for chemical and biological applications.
Roumanie M, et al.
Chemical Engineering Journal, 135, S317-S326 (2008)
The viscosity of triethylamine vapor at low densities and in the saturated vapor phase.
Teske V, et al.
Fluid Phase Equilibria, 293(2), 190-195 (2010)
Kai Morino et al.
Scientific reports, 5, 8953-8953 (2015-05-21)
Well-trained clinicians may be able to provide diagnosis and prognosis from very short biomarker series using information and experience gained from previous patients. Although mathematical methods can potentially help clinicians to predict the progression of diseases, there is no method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service