Skip to Content
Merck
All Photos(1)

Documents

542350

Sigma-Aldrich

1,4-Bis(acryloyl)piperazine

≥99%

Synonym(s):

1,4-Di(acryloyl)piperazine, N,N′-Bis(acryloyl)piperazine, BAP, PDA, Piperazine diacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O2
CAS Number:
Molecular Weight:
194.23
Beilstein:
745431
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99%

mp

91.5-93.5 °C (lit.)

SMILES string

C=CC(=O)N1CCN(CC1)C(=O)C=C

InChI

1S/C10H14N2O2/c1-3-9(13)11-5-7-12(8-6-11)10(14)4-2/h3-4H,1-2,5-8H2

InChI key

YERHJBPPDGHCRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cross-linking monomer for development of polyacrylamide gels.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service