Skip to Content
Merck
All Photos(1)

Key Documents

540447

Sigma-Aldrich

4-Iodo-3-nitrotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(CH3)NO2
CAS Number:
Molecular Weight:
263.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

53-56 °C (lit.)

SMILES string

Cc1ccc(I)c(c1)[N+]([O-])=O

InChI

1S/C7H6INO2/c1-5-2-3-6(8)7(4-5)9(10)11/h2-4H,1H3

InChI key

DKYCDXAZCHMPSJ-UHFFFAOYSA-N

General description

4-Iodo-3-nitrotoluene can be prepared from 4-amino-3-nitrotoluene according to the procedure described by Carlin and Foltz.

Application

4-Iodo-3-nitrotoluene may be used in the preparation of:
  • 3-cyano-4-iodotoluene
  • 4,6′-dimethyl-2 ,2′-dinitrobiphenyl
  • bis(2-nitro-4-methylphenyl) sulfide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The action of sulfur upon para-toluidine in the presence of litharge. Thio-para-toluidine, its constitution and some new derivatives.
Bogert MT and Mandelbaum MR.
Journal of the American Chemical Society, 45(12), 3045-3055 (1923)
A kinetic study of the N-bromosuccinimide bromination of some 4-substituted 3-cyanotoluenes.
Fisher TH and Meierhoefer AW.
The Journal of Organic Chemistry, 43(2), 220-224 (1978)
Ullmann Synthesis of Six Dimethyldinitrobiphenyls and their Reduction to the Corresponding Diaminodimethylbiphenyls1.
Carlin RB and Foltz GE.
Journal of the American Chemical Society, 78(9), 1997-2000 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service