Skip to Content
Merck
All Photos(1)

Documents

538655

Sigma-Aldrich

1-Fluorodecane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F(CH2)9CH3
CAS Number:
Molecular Weight:
160.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.409 (lit.)

bp

185-187 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCF

InChI

1S/C10H21F/c1-2-3-4-5-6-7-8-9-10-11/h2-10H2,1H3

InChI key

LHLRHWJTTUCDQA-UHFFFAOYSA-N

General description

1-Fluorodecane is a 1-fluoroalkane. It undergoes substitution reaction with LiCu(CH3)2, LiCu2(CH3)3, Li2Cu(CH3)3, LiCu(CH3)3, Li2Cu(CH3)3, Li2Cu3(CH3)5 and CH3Li to afford n-undecane (in different yields). 1-Fluorodecane is identified as one of the bio-active compound in the ethanol extract of the stem-bark of Albizia chevalieri and Bridelia ferruginea by GS-MS analysis.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of new organocuprates. 2. Substitution reactions of alkyl, cycloalkyl, and aryl halides with lithium trimethyldicuprate, dilithium trimethylcuprate, and dilithium pentamethyltricuprate.
Ashby EC and Lin JJ.
The Journal of Organic Chemistry, 42(17), 2805-2808 (1977)
GC-MS analysis, acute toxicity and oxidative stress potentials (effects) of Albizia chevalieri extract on juvenile African catfish (Clarias gariepinus).
Ama IU, et al.
Middle-East Journal of Scientific Research 13, 23, 192-199 (2015)
Vapor Pressures and Boiling Points of the 1-Fluoroalkanes, 1-Chloroalkanes, 1-Bromoalkanes, and 1-Iodoalkanes, C1to C20.
Li JCM and Rossini RD.
Journal of Chemical and Engineering Data, 6(2), 268-270 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service