Skip to Content
Merck
All Photos(1)

Documents

53828

Sigma-Aldrich

Hydrazine cyanurate

≥99.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · C3H3N3O3
CAS Number:
Molecular Weight:
161.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (T)

SMILES string

NN.Oc1nc(O)nc(O)n1

InChI

1S/C3H3N3O3.H4N2/c7-1-4-2(8)6-3(9)5-1;1-2/h(H3,4,5,6,7,8,9);1-2H2

InChI key

REWJAYUHYGWDJT-UHFFFAOYSA-N

General description

Hydrazine cyanurate (HC) is a stable source of N2H4 for use in various nitridation reactions. HC can be synthesized from the reaction between cyanuric acid and N2H4.

Application

Hydrazine cyanurate may be used to synthesize anhydrous hydrazine via thermolysis.

Other Notes

Reagent for the preparation of anhydrous hydrazine of high purity

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrazine cyanurate as a nitrogen source for thin nitride film growth
Kropewnicki TJ and Kohl PA
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 16.1 , 139-144 (1998)
Solution behavior of poly (styrene)-B lock-poly (2-vinylpyridine) micelles containing gold nanoparticles
Mossmer S, et al.
Macromolecules, 33.13, 4791- 4798 (2000)
E. Nachbaur et al.
Monatshefte fur Chemie / Chemical Monthly, 102, 1718-1718 (1971)
Development of a 18F-Labeled Tetrazine with Favorable Pharmacokinetics for Bioorthogonal PET Imaging
Denk, Christoph, et al.
Angewandte Chemie (International Edition in English), 53.36, 9655-9659 (2014)
Ruthenium tris (pyrazolyl) borate diazo complexes: Preparation of aryldiazenido, aryldiazene, and hydrazine derivatives
Albertin, Gabriele, et al.
Inorganic Chemistry, 43.14, 4511-4522 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service