Skip to Content
Merck
All Photos(1)

Key Documents

537292

Sigma-Aldrich

1-Chloro-3,5-difluorobenzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3F2
CAS Number:
Molecular Weight:
148.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
95%

Assay

95%

refractive index

n20/D 1.465 (lit.)

bp

111-112 °C (lit.)

density

1.329 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(F)cc(Cl)c1

InChI

1S/C6H3ClF2/c7-4-1-5(8)3-6(9)2-4/h1-3H

InChI key

RFKBODCWHNDUTJ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

85.0 °F - closed cup

Flash Point(C)

29.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service