Skip to Content
Merck
All Photos(1)

Key Documents

536962

Sigma-Aldrich

PAMAM dendrimer solution

25% C₁₂, generation 4, 10 wt. % in methanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$2,611.00

R$2,611.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
5 G
R$2,611.00

About This Item

Empirical Formula (Hill Notation):
C1006H2016N250O156
CAS Number:
Molecular Weight:
20112.35
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

R$2,611.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Product Name

PAMAM-25% C12 dendrimer, generation 4 solution, 10 wt. % in methanol

description

75% amino and 25% [N-(2-hydroxydodecyl)] surface groups

form

solution

mol wt

generation 4

feature

core type ethylenediamine core (2-carbon core)

concentration

10 wt. % in methanol

no. Surface Groups

64

refractive index

n20/D 1.3465

density

0.8 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains ca. 48 primary amino surface groups and 16 [N-(2-hydroxydodecyl)] surface groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service