534773
Hexafluoroacetone deuterate
≥99.5 atom % D
Synonym(s):
Hexafluoro-2-propanone deuterate
About This Item
Recommended Products
isotopic purity
≥99.5 atom % D
form
liquid
technique(s)
NMR: suitable
refractive index
n20/D 1.3 (lit.)
bp
104-108 °C (lit.)
mass shift
M+2
SMILES string
[2H]O[2H].FC(F)(F)C(=O)C(F)(F)F
InChI
1S/C3F6O.H2O/c4-2(5,6)1(10)3(7,8)9;/h;1H2/i/hD2
InChI key
HEBNOKIGWWEWCN-ZSJDYOACSA-N
General description
Application
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service