Skip to Content
Merck
All Photos(1)

Key Documents

534552

Sigma-Aldrich

(R)-(−)-2-Amino-1-hexanol

97%

Synonym(s):

D-Norleucinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(NH2)CH2OH
CAS Number:
Molecular Weight:
117.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −14°, c = 1 in chloroform

bp

216-218 °C (lit.)

mp

40-44 °C (lit.)

SMILES string

CCCC[C@@H](N)CO

InChI

1S/C6H15NO/c1-2-3-4-6(7)5-8/h6,8H,2-5,7H2,1H3/t6-/m1/s1

InChI key

DPEOTCPCYHSVTC-ZCFIWIBFSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

211.0 °F

Flash Point(C)

99.44 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service