Skip to Content
Merck
All Photos(1)

Key Documents

532703

Sigma-Aldrich

1-Bromo-4-chloro-2-fluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)F
CAS Number:
Molecular Weight:
209.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.556 (lit.)

bp

91-92 °C/20 mmHg (lit.)

density

1.678 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(Cl)ccc1Br

InChI

1S/C6H3BrClF/c7-5-2-1-4(8)3-6(5)9/h1-3H

InChI key

FPNVMCMDWZNTEU-UHFFFAOYSA-N

General description

1-Bromo-4-chloro-2-fluorobenzene is a polyhalo substituted benzene. It undergoes Suzuki coupling with 2-cyanoarylboronic esters to form the corresponding biphenyls. These biphenyls are the precursors for synthesizing 6-substituted phenanthridines.† The enthalpy of vaporization at boiling point (467.15K) of 1-bromo-4-chloro-2-fluorobenzene is 40.737kjoule/mol.{4}

Application

1-Bromo-4-chloro-2-fluorobenzene may be used in the preparation of benzonorbornadiene derivative. It may also be used as a starting material in the multi-step synthesis of AZD3264, an IKK2 (inhibitor of nuclear factor κ-B kinase-2) inhibitor.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

198.0 °F - closed cup

Flash Point(C)

92.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of substituted 2-cyanoarylboronic esters.
Lysen M, et al.
The Journal of Organic Chemistry, 71(6), 2518-2520 (2006)
K C Caster et al.
The Journal of organic chemistry, 66(9), 2932-2936 (2001-04-28)
This report details the synthesis of several benzonorbornadienes by Diels--Alder cycloaddition of cyclopentadiene derivatives with substituted benzyne intermediates, which were generated by low-temperature metal--halogen exchange of halobenzenes. General conditions were developed, allowing synthesis of most benzonorbornadienes described herein at the
Exploiting the Differential Reactivities of Halogen Atoms: Development of a Scalable Route to IKK2 Inhibitor AZD3264
Murugan A, et al.
Organic Process Research & Development, 18(5), 646-651 (2014)
Thermophysical Properties of Chemicals and Hydrocarbons, 435-435 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service