Skip to Content
Merck
All Photos(1)

Key Documents

529907

Sigma-Aldrich

(E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester

Synonym(s):

1-[cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)] pentene, 2,2′-[(1Z)-1-Propyl-1,2-ethenediyl]bis[4,4,5,5-tetramethyl-1,3,2-dioxaborolane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H32B2O4
CAS Number:
Molecular Weight:
322.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.455 (lit.)

bp

301-303 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CCC\C(=C\B1OC(C)(C)C(C)(C)O1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C17H32B2O4/c1-10-11-13(19-22-16(6,7)17(8,9)23-19)12-18-20-14(2,3)15(4,5)21-18/h12H,10-11H2,1-9H3/b13-12-

InChI key

MXQDNQSRLNMUOP-SEYXRHQNSA-N

General description

(E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester is an alkenyl boronate ester that can be utilized for Suzuki-Miyaura cross-coupling reaction. Boronate esters are air- and chromatography-stable.

Application

(E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester can be used as a reactant in the stereoselective synthesis of γ-boryl substituted homoallylic alcohols by reacting with aromatic aldehydes via Ru-catalyzed double bond transposition reaction.[1]

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

169.0 °F - closed cup

Flash Point(C)

76.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of γ-Boryl-Substituted Homoallylic Alcohols with anti Stereochemistry Based on a Double-Bond Transposition
Miura T, et al.
Angewandte Chemie (International ed. in English), 58(4), 1138-1142 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service