Skip to Content
Merck
All Photos(1)

Documents

529664

Sigma-Aldrich

Manganese(IV) oxide

≥99.99% trace metals basis

Synonym(s):

Manganese dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

powder and chunks

mp

535 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mn]=O

InChI

1S/Mn.2O

InChI key

NUJOXMJBOLGQSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Y Wang et al.
Journal of colloid and interface science, 380(1), 8-15 (2012-06-02)
Bio-inspired chemical approach has been developed for the surface modification and electrophoretic deposition of manganese dioxide and zirconia nanoparticles, prepared by chemical precipitation methods. Caffeic acid, trans-cinnamic acid, p-coumaric acid, and 2,4-dihydroxycinnamic acid were investigated for the surface modification of
Wen-Hui Kuan et al.
Journal of hazardous materials, 239-240, 152-159 (2012-09-25)
This study examined the reaction of methylene blue (MB) with tunneled manganese oxide pyrolusite regarding pH and reaction time. MB was cleaved through N-demethylation, in which reaction azure B (AB), azure A (AA), azure C (AC), and thionin (TH) were
Michael K L Chu et al.
Lab on a chip, 12(14), 2533-2539 (2012-05-09)
We have developed glucose-responsive implantable microdevices for closed-loop delivery of insulin and conducted in vivo testing of these devices in diabetic rats. The microdevices consist of an albumin-based bioinorganic membrane that utilizes glucose oxidase (GOx), catalase (CAT) and manganese dioxide
Xing Liu et al.
The Analyst, 137(19), 4552-4558 (2012-08-18)
Inorganic nanomaterials that mimic enzymes are fascinating as they potentially have improved properties relative to native enzymes, such as greater resistance to extremes of pH and temperature and lower sensitivity to proteases. Although many artificial enzymes have been investigated, searching
Highly stretchable alkaline batteries based on an embedded conductive fabric.
Abhinav M Gaikwad et al.
Advanced materials (Deerfield Beach, Fla.), 24(37), 5071-5076 (2012-07-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service