Skip to Content
Merck
All Photos(1)

Key Documents

528951

Sigma-Aldrich

Methyl 2-fluorobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CO2CH3
CAS Number:
Molecular Weight:
154.14
Beilstein:
1862493
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.502 (lit.)

bp

109-110 °C/35 mmHg (lit.)

density

1.21 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1F

InChI

1S/C8H7FO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,1H3

InChI key

QAFJIJWLEBLXHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 2-fluorobenzoate is an ortho-halogen-substituted methyl benzoate ester.[1] It reacts with hydrazide to afford 2-fluorobenzoic hydrazide.[2] Methyl 2-fluorobenzoate undergoes enzymatic dihydroxylation via the whole-cell fermentation in the presence of Escherichia coli JM109 (pDTG601A) to afford a diol.[1]

Application

Methyl 2-fluorobenzoate may be used to synthesize 2-fluoro-α-methylstyrene[3] and 2-fluorophenyldiphenylmethanol.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.8 °F - closed cup

Flash Point(C)

93.80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aromatic Fluorine Compounds. X. The 2, 3-and 2, 6-Difluoropyridines1.
Finger GC, et al.
The Journal of Organic Chemistry, 27(11), 3965-3396 (1962)
Vladislav Semak et al.
Organic & biomolecular chemistry, 10(22), 4407-4416 (2012-05-09)
A series of ortho-, meta-, and para- halogen-substituted methyl benzoate esters was subjected to enzymatic dihydroxylation via the whole-cell fermentation with E. coli JM109 (pDTG601A). Only ortho-substituted benzoates were metabolized. Methyl 2-fluorobenzoate yielded one diol regioselectively whereas methyl 2-chloro-, methyl
Monomers and Polymers. II. a-Methylstyrenes and the Steric Hindrance of ortho-Substituents1.
Bachman GB and Finholt RW.
Journal of the American Chemical Society, 70(2), 622-624 (1948)
The intramolecular C-F? HO hydrogen bond of 2-fluorophenyldiphenylmethanol.
Takemura H, et al.
New. J. Chem., 33(10), 2004-2006 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service